missing translation for 'onlineSavingsMsg'
Learn More

ABL1/ABL2 (phospho Y393/Y429), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic phosphopeptide of ABL1/ABL2.

Brand:  Abnova PAB12190.100uL

Product Code. 16273240

  • £349.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: DTYpTA
Specifications

Specifications

ABL1/ABL2
Polyclonal
Unconjugated
Immunohistochemistry (1:50-1:100) Immunofluorescence (1:100-1:200) The optimal working dilution should be determined by the end user.
P00519 (Gene ID : 25);P42684 (Gene ID : 27)
ABL1
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y393/Y429 of human ABL1/ABL2.
100 μL
Primary
ABL1/2 (phospho-Tyr393/429) Antibody detects endogenous levels of ABL1/2 only when phosphorylated at tyrosine393/429.
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Immunofluorescence, Immunohistochemistry (PFA fixed)
1 mg/mL
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ABL1/ABL2.
In PBS (without mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
ABL/JTK7/bcr/abl/c-ABL/p150/v-abl
Rabbit
Immunoaffinity purification
RUO
25/27
Human, Mouse
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only