missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Rabbit actin native Protein from rabbit muscle
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Specifications
Specifications
| For Use With (Application) | Antibody Production, SDS-PAGE |
| Formulation | In 10mM Tris/HCl, pH 8.0, 0.2mM CaCl, |
| Molecular Weight (g/mol) | 43kDa |
| Name | Actin (Rabbit) Native Protein |
| Preparation Method | Native protein purified from rabbit muscle |
| Quantity | 100 μg |
| Storage Requirements | Store at 4°C. This product is stable for at least 3 months. After reconstitution, store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Regulatory Status | RUO |
| Common Name | ACTIN |
| Species | Rabbit |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction