missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB9B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | RAB9B |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16878204
|
Proteintech
18152-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16868204
|
Proteintech
18152-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ras-related protein Rab-9A, also called RAB9, is a protein that in humans is encoded by the RAB9A gene. This gene is mapped to Xp22.2. RAB9 has been localized to components of the endocytic/exocytic pathway and has been implicated in recycling of membrane receptors. It has been found that downregulation of RAB9A gene expression in HeLa cells induced severe cell vacuolation. RAB9A GTPase is directly bound by TIP47 in its active, GTP-bound conformation. Moreover, RAB9A increases the affinity of TIP47 for its cargo. What′s more, this gene may involved in the transport of proteins between the endosomes and the trans Golgi network. RAB9A has been shown to interact with RABEPK and TIP47.Specifications
| RAB9B | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| Q8BHH2, Q9NP90 | |
| 319642, 367915, 51209 | |
| RAB9B Fusion Protein Ag12848 | |
| Primary | |
| -20°C | |
| RAB9B |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Rab 9 like protein, Rab 9L, RAB9B, RAB9L, Ras related protein Rab 9B | |
| Rab9b | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title