missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB9B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570252
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 79%; Zebrafish: 79%.
Rab9b may be involved in the transport of proteins between the endosomes and the trans Golgi network.
Specifications
| RAB9B | |
| Polyclonal | |
| Unconjugated | |
| RAB9B | |
| 9330195C02Rik; Rab9b; RAB9B, member RAS oncogene family; RAB9L; Rab-9L; Rab-9-like protein; Ras-related protein Rab-9B; RGD1562951; si:dkey-169h1.8; TRAB9 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 319642 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8BHH2 | |
| RAB9B | |
| synthetic peptide directed towards the middle region of human Rab9 (aa 58-107). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction