missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB9B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA568308
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
This gene encodes a member of a subfamily of RAS small guanosine triphosphate (GTP)-binding proteins that regulate membrane trafficking. The encoded protein may be involved in endosome-to-Golgi transport.
Specifications
| RAB9B | |
| Polyclonal | |
| Unconjugated | |
| RAB9B | |
| RAB9-like protein; rab-9-like protein; ras-related protein Rab-9B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 319642, 367915, 51209 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NP90, Q8BHH2 | |
| RAB9B | |
| Synthesized peptide derived from human Rab9. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion