missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB9 Recombinant Rabbit Monoclonal Antibody (23GB1940), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553181
This item is not returnable.
View return policy
Description
Rab proteins are a family of Ras-like GTPases involved in intracellular compartment protein transport. Different members of the 40+ member rab family are responsible for docking and fusion of transport vesicles between different compartments within the cell. Rab 9 is required for trafficking mannose 6-phosphate receptor between the late endosome to trans-Golgi network (TGN). By facilitating receptor transport, rab 9 enables cells to efficiently recycle important cellular trafficking components. It is functionally necessary for rab 9, like other rab family members, to be prenylated by two 20-carbon geranylgeranyl groups at the C-terminus. Most prenylated rab 9 is membrane bound, however, 10-20% of the cellular pool of rab 9 is bound to GDP dissociation inhibitor-alpha (GDI-alpha) in the cytosol. GDI recycles prenylated, GDP bound rab 9 from their fusion targets back to their membranes of origin.
Specifications
| RAB9 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P51151, Q99P75, Q9R0M6 | |
| Rab9, Rab9a | |
| A synthesized peptide derived from human Rab9 (30-74AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1940 | |
| Unconjugated | |
| Rab9a | |
| 2410064E05Rik; AI195561; rab9; rab9 GTP-binding protein; RAB9, member RAS oncogene family; Rab9a; rab-9A; RAB9A, member RAS oncogene family; RAB9-like protein; ras-related protein Rab-9A; Sid 99; Sid6061p; Sid99 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 56382, 84589, 9367 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction