missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAB7A Polyclonal Antibody
GREENER_CHOICE

Product Code. 16586871
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16586871 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16586871 Supplier Invitrogen™ Supplier No. PA588755

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Samples: A-431, HeLa, Mouse brain, Rat brain; Cellular Location: Cytoplasmic side, Cytoplasmic vesicle, Late endosome membrane, Lipid droplet, Lysosome membrane, Melanosome membrane, Peripheral membrane protein, autophagosome membrane, phagosome membrane Immunogen sequence: DSWRDEFLIQ ASPRDPENFP FVVLGNKIDL ENRQVATKRA QAWCYSKNNI PYFETSAKEA INVEQAFQTI ARNALKQETE VELYNEFPEP IKLDKNDRAK ASAESCSC.

RAB family members are small, RAS-related GTP-binding proteins that are important regulators of vesicular transport. Each RAB protein targets multiple proteins that act in exocytic / endocytic pathways. This gene encodes a RAB family member that regulates vesicle traffic in the late endosomes and also from late endosomes to lysosomes. This encoded protein is also involved in the cellular vacuolation of the VacA cytotoxin of Helicobacter pylori. Mutations at highly conserved amino acid residues in this gene have caused some forms of Charcot-Marie-Tooth (CMT) type 2 neuropathies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB7A
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene RAB7A
Gene Accession No. P09527, P51149, P51150
Gene Alias GTP-binding protein (rab7); hypothetical protein LOC509970; PRO2706; RAB7; rab-7; RAB7, member RAS oncogene family; Rab7a; RAB7A, member RAS oncogene family; Ras-associated protein RAB7; ras-related protein BRL-RAS; Ras-related protein p23; ras-related protein Rab-7a
Gene Symbols Rab7, RAB7A
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110315128, 19349, 29448, 7879
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.