missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB7A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578237
This item is not returnable.
View return policy
Description
Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Rat (99%), Xenopus laevis (95%), Dog (100%), Rabbit (97%), Sheep (99%), Bovine (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RAB family members are small, RAS-related GTP-binding proteins that are important regulators of vesicular transport. Each RAB protein targets multiple proteins that act in exocytic / endocytic pathways. This gene encodes a RAB family member that regulates vesicle traffic in the late endosomes and also from late endosomes to lysosomes. This encoded protein is also involved in the cellular vacuolation of the VacA cytotoxin of Helicobacter pylori. Mutations at highly conserved amino acid residues in this gene have caused some forms of Charcot-Marie-Tooth (CMT) type 2 neuropathies.
Specifications
| RAB7A | |
| Polyclonal | |
| Unconjugated | |
| RAB7A | |
| GTP-binding protein (rab7); hypothetical protein LOC509970; PRO2706; RAB7; rab-7; RAB7, member RAS oncogene family; Rab7a; RAB7A, member RAS oncogene family; Ras-associated protein RAB7; ras-related protein BRL-RAS; Ras-related protein p23; ras-related protein Rab-7a | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 110315128, 19349, 7879 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.4 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P51149, P51150 | |
| Rab7, RAB7A | |
| Recombinant protein encompassing a sequence within the center region of human RAB7A. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction