missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB7 Recombinant Rabbit Monoclonal Antibody (24GB895)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557666
This item is not returnable.
View return policy
Description
RAB7 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
RAB family members are small, RAS-related GTP-binding proteins that are important regulators of vesicular transport. Each RAB protein targets multiple proteins that act in exocytic / endocytic pathways. This gene encodes a RAB family member that regulates vesicle traffic in the late endosomes and also from late endosomes to lysosomes. This encoded protein is also involved in the cellular vacuolation of the VacA cytotoxin of Helicobacter pylori. Mutations at highly conserved amino acid residues in this gene have caused some forms of Charcot-Marie-Tooth (CMT) type 2 neuropathies.
Specifications
| RAB7 | |
| Recombinant Monoclonal | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P09527, P51149, P51150 | |
| Rab7, RAB7A | |
| A synthesized peptide derived from human RAB7 (aa 150-207). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB895 | |
| Unconjugated | |
| RAB7A | |
| GTP-binding protein (rab7); hypothetical protein LOC509970; PRO2706; RAB7; rab-7; RAB7, member RAS oncogene family; Rab7a; RAB7A, member RAS oncogene family; Ras-associated protein RAB7; ras-related protein BRL-RAS; Ras-related protein p23; ras-related protein Rab-7a | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 110315128, 19349, 29448, 7879 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction