missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB6C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101829
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RAB6C.
May be involved in the regulation of centrosome duplication and cell cycle progression.
Specifications
| RAB6C | |
| Polyclonal | |
| Unconjugated | |
| RAB6C | |
| RAB6C; RAB6C, member RAS oncogene family; rab6-like protein WTH3; ras-related protein Rab-6C; WTH3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 84084 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9H0N0 | |
| RAB6C | |
| A synthesized peptide derived from human RAB6C(Accession Q9H0N0), corresponding to amino acid residues K158-C208. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction