missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAB6B Monoclonal Antibody (6D12E4)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA531882

Product Code. 16315314

  • £433.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA5-31882 has been tested in indirect ELISA.

The RAB6B prodtein seems to have a role in retrograde membrane traffic at the level of the Golgi complex. It may also function in retrograde transport in neuronal cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RAB6B
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
Q9NRW1
Rab6b
Purified recombinant fragment of human Rab6b (AA: 95-208) expressed in E. coli.
100 μL
Primary
Human, Rat
Antibody
IgG1
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
6D12E4
Unconjugated
Rab6b
9630015C03; AI844641; C330006L04Rik; D9Bwg0185e; Rab6b; RAB6B, member RAS oncogene family; ras-related protein Rab-6B; small GTPase RAB6B; small GTP-binding protein
Mouse
Protein G
RUO
363123, 51560
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.