missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB6A Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-25946C
This item is not returnable.
View return policy
Description
RAB6A Polyclonal antibody specifically detects RAB6A in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| RAB6A | |
| Polyclonal | |
| Western Blot, ELISA, Immunoprecipitation | |
| GTP-binding protein RAB6A', GTP-binding protein RAB6B, Oncogene RAB6, Rab GTPase, Rab-6, RAB6, member RAS oncogene family, RAB6A', Rab6A variant 3, RAB6A, member RAS oncogene family, RAB6B, RAB6rab-6, ras-related protein Rab-6A | |
| Rabbits immunized with purified, recombinant Human RAB6A (Accession#: P20340-2, Met1-Cys208) | |
| 0.1 mL | |
| Signal Transduction | |
| 5870 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction