missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB5C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110681
This item is not returnable.
View return policy
Description
Immunogen sequence: NSLLFMETSA KTAMNVNEIF MAIAKKLPKN EPQNATGAPG RNRGVDLQEN NPASRSQCCS N Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 95%.
The Ras-related superfamily of guanine nucleotide binding proteins, which includes the R-Ras, Rap, Ral/Rec and Rho/Rab subfamilies, exhibit 30-60% homology with Ras p21. Accumulating data suggests an important role for Rab proteins, either in endocytosis or in biosynthetic protein transport. The transport of newly synthesized proteins from the endoplasmic reticulum to various stacks of the Golgi complex and to secretory vesicles involves at each stage the movement of carrier vesicles, a process that appears to involve Rab protein function. The possibility that Rab proteins might also direct the exocytosis from secretory vesicles to the plasma membrane is supported by the observation that in yeast, the SEC4 protein, which is 40% homologous to Rab proteins, is associated with secretory vesicles.
Specifications
| RAB5C | |
| Polyclonal | |
| Unconjugated | |
| RAB5C | |
| AI326010; L1880; RAB; RAB5B; RAB5B, member RAS oncogene family; RAB5C; Rab5c protein; RAB5C, member of RAS oncogene family; RAB5C, member RAS oncogene family; RAB5C, member RAS oncogene family isoform b; RAB5CL; RAB5L; RABL; Ras-related protein Rab-5C; small GTPase; wu:fb98b08; wu:fi16g01 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5878 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P51148 | |
| RAB5C | |
| Recombinant protein fragment of Human RAB5C (Product #RP-102038) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction