missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Rab5b Polyclonal antibody specifically detects Rab5b in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Rab5b |
| Applications | Western Blot |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Rab5b. |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?