missing translation for 'onlineSavingsMsg'
Learn More

Rab5a Rabbit anti-Human, Mouse, Rat, Clone: 0K5J1, Novus Biologicals™

Product Code. 18382976 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382976 20 μg 20µL
18382594 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18382976 Supplier Novus Biologicals Supplier No. NBP31544220UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Rab5a Monoclonal antibody specifically detects Rab5a in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rab5a
Applications Western Blot
Classification Monoclonal
Clone 0K5J1
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias RAB5A, member RAS oncogene family, RAB5RAS-associated protein RAB5A, ras-related protein Rab-5A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 116-215 of human Rab5aA (P20339).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Endosome Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5868
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.