missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab5a Antibody (3A4), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04340AF532
This item is not returnable.
View return policy
Description
Rab5a Monoclonal specifically detects Rab5a in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Rab5a | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| RAB5A | |
| Recombinant human Rab5a (1-215aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG2b κ |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 3A4 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| RAB5A, member RAS oncogene family, RAB5RAS-associated protein RAB5A, ras-related protein Rab-5A | |
| Mouse | |
| Protein G purified | |
| Endosome Markers, Signal Transduction | |
| 5868.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction