missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB40A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104078
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RAB40A.
This gene encodes a member of the Rab40 subfamily of Rab small GTP-binding proteins that contains a C-terminal suppressors of cytokine signaling box.
Specifications
| RAB40A | |
| Polyclonal | |
| Unconjugated | |
| RAB40A | |
| protein Rar-2; RAB40A; RAB40A, member RAS oncogene family; RAR2; RAR2A; Ras-related protein Rab-40A; SOCS box containing protein RAR2A; SOCS box-containing protein RAR2A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 142684 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8WXH6 | |
| RAB40A | |
| A synthesized peptide derived from human RAB40A(Accession Q8WXH6), corresponding to amino acid residues T244-S277. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction