missing translation for 'onlineSavingsMsg'
Learn More

Rab4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18378003
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378003 100 μg 100µL
18378604 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18378003 Supplier Bio-Techne Supplier No. NBP318233100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Rab4 Polyclonal antibody specifically detects Rab4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rab4
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:150
Formulation PBS pH 7.4, 50% glycerol
Gene Alias Oncogene RAB4, RAB4, member RAS oncogene family, RAB4A, member RAS oncogene family, RAB4HRES-1/RAB4, ras-related protein Rab-4A
Host Species Rabbit
Immunogen C-terminal peptide from human Rab4
Purification Method Peptide affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5867
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.