Learn More
Invitrogen™ Rab4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA3912
Description
PA3-912 detects Rab4 from human and mouse samples. This antibody does not detect other Rab isoforms (Rab1, Rab3, Rab5, or Rab6). PA3-912 has been successfully used in Western blot and immunofluorescence procedures. By Western blot, this antibody detects a ~24 kDa band representing Rab4 from transfected HeLa cells. The PA3-912 immunogen is purified recombinant human Rab4 protein.
Protein transport. Probably involved in vesicular traffic.
Specifications
| Rab4 | |
| Polyclonal | |
| Unconjugated | |
| Rab4a | |
| AI848268; HRES1; HRES-1; HRES-1/RAB4; HTLV-1 related endogenous sequence; RAB4; Rab4a; RAB4A, member RAS oncogene family; ras-related protein Rab-4A; Ras-related small GTP binding protein 4 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Antibody | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Conc. Not Determined | |
| Whole serum with 0.05% sodium azide | |
| P05714, P20338, P56371 | |
| Rab4a | |
| Purified, recombinant human Rab4 protein. | |
| RUO | |
| 19341, 25532, 479210, 5867 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.