Learn More
Invitrogen™ Rab4 Monoclonal Antibody (4E11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517161
Description
MA5-17161 targets RAB4A in FACS, IHC, indirect ELISA, and WB applications and shows reactivity with Human, Mouse, and Non-human primate samples. The MA5-17161 immunogen is purified recombinant fragment of human RAB4A expressed in E. Coli. MA5-17161 detects RAB4A which has a predicted molecular weight of approximately 23.8kDa.
Protein transport. Probably involved in vesicular traffic.
Specifications
| Rab4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P20338, P56371 | |
| Rab4a | |
| Purified recombinant fragment of human RAB4A expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 4E11 | |
| Unconjugated | |
| Rab4a | |
| AI848268; HRES1; HRES-1; HRES-1/RAB4; HTLV-1 related endogenous sequence; RAB4; Rab4a; RAB4A, member RAS oncogene family; ras-related protein Rab-4A; Ras-related small GTP binding protein 4 | |
| Mouse | |
| Protein G | |
| RUO | |
| 19341, 5867 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.