missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab4 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18233H
This item is not returnable.
View return policy
Description
Rab4 Polyclonal antibody specifically detects Rab4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Rab4 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| Oncogene RAB4, RAB4, member RAS oncogene family, RAB4A, member RAS oncogene family, RAB4HRES-1/RAB4, ras-related protein Rab-4A | |
| C-terminal peptide from human Rab4 | |
| 0.1 mL | |
| Signal Transduction | |
| 5867 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| HRP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction