missing translation for 'onlineSavingsMsg'
Learn More

Rab4 Antibody (1C10), Novus Biologicals™

Product Code. 18369978 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369978 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369978 Supplier Novus Biologicals Supplier No. H00005867M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Rab4 Monoclonal antibody specifically detects Rab4 in Human samples. It is validated for Western Blot, ELISA, ELISA, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rab4
Applications Western Blot, ELISA, Sandwich ELISA, KnockDown
Classification Monoclonal
Clone 1C10
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA, Knockdown Validated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH04309
Gene Alias Oncogene RAB4, RAB4, member RAS oncogene family, RAB4A, member RAS oncogene family, RAB4HRES-1/RAB4, ras-related protein Rab-4A
Host Species Mouse
Immunogen RAB4A (AAH04309, 1 a.a. ~ 218 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSQTAMSETYDFLFKFLVIGNAGTGKSCLLHQFIEKKFKDDSNHTIGVEFGSKIINVGGKYVKLQIWDTAGQERFRSVTRSYYRGAAGALLVYDITSRETYNALTNWLTDARMLASQNIVIILCGNKKDLDADREVTFLEASRFAQENELMFLETSALTGENVEEAFVQCARKILNKIESGELDPERMGSGIQYGDAALRQLRSPRRAQAPNAQECGC
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5867
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.