missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB3IP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£342.00
Specifications
| Antigen | RAB3IP |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:500-1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
RAB3IP Polyclonal specifically detects RAB3IP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| RAB3IP | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FLJ14660, FLJ22548, MGC71495, RAB3A interacting protein (rabin3), rab-3A-interacting protein, Rab3A-interacting protein, rabin-3, RABIN3, SSX2 interacting protein, SSX2-interacting protein | |
| RAB3IP | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:500-1:1000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q96QF0 | |
| 117177 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:AEISSISFHVTDPAPCSTSGVTAGLTKLTTRKDNYNAEREFLQGATITEACDGSDDIFGLSTDSLSRLRSPSVLEVREKGYERLKEELAKAQ | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| 53 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title