missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB3GAP2 Polyclonal Antibody

RAB3GAP2 Polyclonal Antibody
Brand: Thermo Scientific PA555574
This item is not returnable.
View return policy
Description
RAB3GAP2 Polyclonal Antibody for IHC
Specifications
| RAB3GAP2 | |
| Polyclonal | |
| Unconjugated | |
| RAB3GAP2 | |
| 1110059F07Rik; 2010002H18Rik; 5830469C09; AI851069; AW743433; KIAA0839; mKIAA0839; p150; RAB3 GTPase activating non-catalytic protein subunit 2; RAB3 GTPase activating protein subunit 2; RAB3 GTPase activating protein subunit 2 (non-catalytic); Rab3 GTPase-activating protein 150 kDa subunit; Rab3 GTPase-activating protein non-catalytic subunit; rab3-GAP p150; Rab3-GAP regulatory subunit; RAB3GAP150; RAB3-GAP150; RAB3GAP2; RGAP-iso; RGD1311518; SPG69; WARBM2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25782 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H2M9 | |
| RAB3GAP2 | |
| Recombinant protein corresponding to Human RAB3GAP2. Recombinant protein control fragment (Product #RP-94142). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction