missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB3GAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556952
This item is not returnable.
View return policy
Description
Immunogen sequence: PISELHLATT WPHLTEGIIV DNDVYSDLDP IQAPHWSVRV RKAENPQCLL GDFVTEFFKI CRRKESTDEI LGRSAFEEEG KETADITHAL SK Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.
RAB3GAP1, is a 981 amino acid protein that belongs to the Rab3-GAP catalytic subunit family. Rab 3 GAP p130 converts active RAB3-GTP to the inactive form RAB3-GDP, and is required for normal eye and brain development. Defects in Rab 3 GAP p130 are the cause of Warburg micro syndrome 1 (WARBM1). WARBM1 is a severe autosomal recessive disorder characterized by developmental abnormalities of the eye and central nervous system and by microgenitalia. The Rab 3 GAP p130 protein may participate in neurodevelopmental processes such as proliferation, migration and differentiation before synapse formation, and non-synaptic vesicular release of neurotransmitters.
Specifications
| RAB3GAP1 | |
| Polyclonal | |
| Unconjugated | |
| RAB3GAP1 | |
| 1700003B17Rik; 4732493F09Rik; AL117896; KIAA0066; P130; Rab 3 GAP p130; Rab3 GAP p130; RAB3 GTPase activating protein catalytic subunit 1; RAB3 GTPase activating protein subunit 1; RAB3 GTPase activating protein subunit 1 (catalytic); RAB3 GTPase-activating protein 130 kDa subunit; Rab3 GTPase-activating protein catalytic subunit; RAB3GAP; Rab3-GAP; Rab3-GAP p130; Rab3gap1; RAB3GAP130; RGD1306487; WARBM1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22930 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15042 | |
| RAB3GAP1 | |
| Recombinant protein corresponding to Human RAB3GAP1. Recombinant protein control fragment (Product #RP-102619). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction