missing translation for 'onlineSavingsMsg'
Learn More

RAB3GAP1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18404433 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404433 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404433 Supplier Novus Biologicals Supplier No. NBP119154JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RAB3GAP1 Polyclonal antibody specifically detects RAB3GAP1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB3GAP1
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias KIAA0066P130, RAB3 GTPase activating protein subunit 1 (catalytic), RAB3 GTPase-activating protein 130 kDa subunit, rab3 GTPase-activating protein catalytic subunit, Rab3-GAP, Rab3-GAP p130, RAB3GAPDKFZp434A012, WARBM1RAB3GAP130
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 350 and 400 of human RAB3 GTPase activating protein subunit 1 using the numbering given in entry NP_036365.1 (GeneID 22930).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, GPCR, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 22930
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.