Learn More
RAB3D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12320-1-AP-20UL
 View more versions of this product
Product Code. 16899963
  • £135.00 / 20µL
Description
Rab proteins are low-molecular-weight GTP-binding proteins that form the largest branch of the Ras superfamily of GTPases. Located on the cytoplasmic face of organelles and vesicles, rab proteins are involved in intracellular membrane fusion reactions. Three membrane proteins, synaptosomal associated protein of 25 kDa (SNAP-25), synaptobrevin, and syntaxin, form the core of a ubiquitous membrane fusion machine that interacts with the soluble proteins N-ethylmaleimide-sensitive factor (NSF) and a-SNAP. Rab proteins, in coordination with the core fusion machinery and Munc-18, help to mediate vesicle docking and fusion. There exist over 40 rab proteins that have been described in mammals. Rab 3D has been shown to be present, in part, in the tongue, pancreas, and liver.
Specifications
Specifications
RAB3D
Polyclonal
Unconjugated
RAB3D
D2 2, GOV, RAB16, RAB3D, RAD3D, Ras related protein Rab 3D
Rabbit
Antigen Affinity Chromatography
RUO
140665, 19340, 9545
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.49 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95716, P35276, Q63942
RAB3D
RAB3D Fusion Protein Ag2973
20 μL
Primary
Rat, Human, Mouse