missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Rab3D Monoclonal antibody specifically detects Rab3D in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Rab3D |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 5000000000 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000 |
| Formulation | PBS pH 7.4, 50% glycerol |
| Gene Alias | D2-2, GOVras-related protein Rab-3D, RAB16glioblastoma overexpressed, RAB3D, member RAS oncogene family, RAD3DRab3D upregulated with myeloid differentiation |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?