missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB3a Monoclonal Antibody (1531CT562.14.57)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537684
This item is not returnable.
View return policy
Description
RAB3a Monoclonal Antibody for Western Blot, ICC/IF, Flow
Rab proteins belong to the Ras small GTPase superfamily. The GTP-binding Rab3 subfamily has 4 known isoforms: 3A, 3B, 3C, and 3D. These proteins are expressed in highest concentrations in brain and endocrine tissues and involved in regulated exocytosis of hormones and neurotransmitters. Specifically, Rab 3s are thought to play a key role in presynaptic vesicle trafficking and priming.
Specifications
| RAB3a | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P20336, P63011, P63012 | |
| RAB3A | |
| RAB3a recombinant protein. | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 1531CT562.14.57 | |
| Unconjugated | |
| RAB3A | |
| C18A3.6; RAB3; Rab3a; RAB3A, member RAS oncogene family; RAS-associated protein RAB3A; ras-related protein Rab-3A; ras-related protein Rab-3A-like protein; Ras-related small GTP binding protein 3A | |
| Mouse | |
| Protein G | |
| RUO | |
| 19339, 25531, 5864 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction