Learn More
RAB38 Mouse, anti-Human, Clone: 3A10C5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68603-1-IG-20UL
 View more versions of this product
Product Code. 18095285
  • £135.00 / 20µL
Description
The RAB38 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAB38. This antibody recognizes human antigen. The RAB38 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
RAB38
3A10C5
Melanoma antigen NY MEL 1, NY MEL 1, RAB38, Ras related protein Rab 38, rrGTPbp
20 μL
23682
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human
150 kDa
IgG2a