Learn More
RAB38 Mouse, anti-Human, Clone: 3A10C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68603-1-IG-150UL
This item is not returnable.
View return policy
Description
The RAB38 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAB38. This antibody recognizes human antigen. The RAB38 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| RAB38 | |
| 3A10C5 | |
| Melanoma antigen NY MEL 1, NY MEL 1, RAB38, Ras related protein Rab 38, rrGTPbp | |
| 150 μL | |
| 23682 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG2a |
Product Content Correction
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu