missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB37 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13051-1-AP-150UL
This item is not returnable.
View return policy
Description
Rab proteins are low molecular mass GTPases that are critical regulators of vesicle trafficking. For additional background information on Rab proteins.Specifications
| RAB37 | |
| Polyclonal | |
| Unconjugated | |
| RAB37 | |
| RAB37, Ras related protein Rab 37 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 326624, 58222 | |
| -20°C | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96AX2, Q9JKM7 | |
| RAB37 | |
| RAB37 Fusion Protein Ag3717 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction