missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB37 Antibody (OTI2F1), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73772FR
This item is not returnable.
View return policy
Description
RAB37 Monoclonal specifically detects RAB37 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| RAB37 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| RAB37 | |
| Full length recombinant protein of human RAB37 (NP_783865) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2F1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| FLJ32507, member of RAS oncogene family, RAB37, member RAS oncogene family, ras-related protein Rab-37 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 326624.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction