missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAB36 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15524301
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15524301 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15524301 Supplier Invitrogen™ Supplier No. PA537207

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 37 kDa. Purity is >95% by SDS-PAGE.

Protein transport. Probably involved in vesicular traffic. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB36
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene RAB36
Gene Accession No. O95755, Q8CAM5
Gene Alias 6330593L16Rik; AW047545; Rab36; RAB36, member RAS oncogene family; ras-related protein Rab-36; small GTP-binding protein Rab36
Gene Symbols RAB36
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues in human RAB36.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 690407, 76877, 9609
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.