missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RAB33A Polyclonal antibody specifically detects RAB33A in Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | RAB33A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 50% glycerol, pH7.3 |
| Gene Alias | MGC1488, RAB33A, member RAS oncogene family, RabS10, ras-related protein Rab-33A, Small GTP-binding protein S10 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 118-237 of human RAB33A (NP_004785.1). |
| Purification Method | Affinity purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?