missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB32 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10999-1-AP-150UL
This item is not returnable.
View return policy
Description
RAB32 acts as an A-kinase anchoring protein by binding to the type II regulatory subunit of protein kinase A and anchoring it to the mitochondrion. Also involved in synchronization of mitochondrial fission. Plays a role in the maturation of phagosomes that engulf pathogens, such as S.aureus and M.tuberculosis.Specifications
| RAB32 | |
| Polyclonal | |
| Unconjugated | |
| RAB32 | |
| RAB32, Ras related protein Rab 32 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10981 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13637 | |
| RAB32 | |
| RAB32 Fusion Protein Ag1450 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction