missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB2B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31631
This item is not returnable.
View return policy
Description
RAB2B Polyclonal specifically detects RAB2B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| RAB2B | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris-Glycine (pH7), 10% Glycerol with 0.01% Thimerosal | |
| RAB2B | |
| Full length human RAB2B Recombinant protein. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.83mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| FLJ14824, member RAB2B, RAB2B, member RAS oncogene family, ras-related protein Rab-2B | |
| Rabbit | |
| 24 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 84932 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction