missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB2A Mouse anti-Human, Unconjugated, Clone: 3E1B10, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | RAB2A |
|---|---|
| Clone | 3E1B10 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16860166
|
Proteintech
67501-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16850166
|
Proteintech
67501-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RAB2A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAB2A. This antibody recognizes human, mouse, rat antigen. The RAB2A antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| RAB2A | |
| 500 μg/mL | |
| Monoclonal | |
| Mouse | |
| 5862 | |
| Primary |
| 3E1B10 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG2b |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title