missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB26 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14284-1-AP-20UL
This item is not returnable.
View return policy
Description
Members of the RAB protein family, including RAB26, are important regulators of vesicular fusion and trafficking. The RAB family of small G proteins regulates intercellular vesicle trafficking, including exocytosis, endocytosis, and recycling (summary by Seki et al., 2000 ).Specifications
| RAB26 | |
| Polyclonal | |
| Unconjugated | |
| RAB26 | |
| RAB26, Ras related protein Rab 26, V46133 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25837, 328778 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q504M8, Q9ULW5 | |
| Rab26 | |
| RAB26 Fusion Protein Ag5581 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction