missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAB26 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13405807
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13405807 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13405807 Supplier Invitrogen™ Supplier No. PA531287

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG. Predicted reactivity: Human (99%), Mouse (91%), Rat (91%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Members of the RAB protein family, including RAB26, are important regulators of vesicular fusion and trafficking. The RAB family of small G proteins regulates intercellular vesicle trafficking, including exocytosis, endocytosis, and recycling (summary by Seki et al., 2000 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB26
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene RAB26
Gene Accession No. P51156, Q9ULW5
Gene Alias A830020M03Rik; RAB26; RAB26, member RAS oncogene family; Ras-related oncogene protein; ras-related protein Rab-26; V46133
Gene Symbols RAB26
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RAB26. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171111, 25837
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.