missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab25 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | Rab25 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Rab25 Polyclonal specifically detects Rab25 in Human samples. It is validated for Western Blot.Specifications
| Rab25 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CATX8, CATX-8RAB11C, RAB25, member RAS oncogene family, ras-related protein Rab-25 | |
| The immunogen is a synthetic peptide directed towards the N terminal region of human Rab25 (NP_065120.2). Peptide sequence GNGTEEDYNFVFKVVLIGESGVGKTNLLSRFTRNEFSHDSRTTIGVEFST | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 57111 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title