missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598105
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
RAB25 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
RAB proteins, such as RAB25, are members of the RAS superfamily of small GTPases that are involved in membrane trafficking. Members of the RAB11 subfamily, including RAB25, control the return of internalized membrane-associated moieties to the cell surface.
Spécification
| RAB25 | |
| Polyclonal | |
| Unconjugated | |
| RAB25 | |
| AV012978; CATX8; CATX-8; RAB11C; RAB25; RAB25, member RAS oncogene family; ras-related protein Rab-25 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 53868, 57111 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P57735, Q9WTL2 | |
| RAB25 | |
| Recombinant Human Ras-related protein Rab-25 protein (1-210AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu