missing translation for 'onlineSavingsMsg'
Learn More

Rab25 Antibody [Allophycocyanin], Novus Biologicals™

Product Code. 30737631 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30737631 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737631 Supplier Novus Biologicals Supplier No. AF6605A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Rab25 Polyclonal antibody specifically detects Rab25 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rab25
Applications Western Blot
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias CATX8, CATX-8RAB11C, RAB25, member RAS oncogene family, ras-related protein Rab-25
Host Species Goat
Immunogen E. coli-derived recombinant human RAB25, Met1-Ala195, Accession # P57735
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57111
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.