missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab25 Antibody (3F12F3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-28845-0.025ml
This item is not returnable.
View return policy
Description
Rab25 Monoclonal specifically detects Rab25 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Rab25 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| CATX8, CATX-8RAB11C, RAB25, member RAS oncogene family, ras-related protein Rab-25 | |
| Mouse | |
| Protein A or G purified | |
| Primary | |
| Specific for Rab25. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2a |
| Western Blot | |
| 3F12F3 | |
| Unconjugated | |
| PBS with 0.03% Sodium Azide | |
| RAB25 | |
| Purified recombinant fragment of Rab25 expressed in E. Coli. | |
| 0.025 mL | |
| 57111 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction