missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB24 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145189
This item is not returnable.
View return policy
Description
Immunogen sequence: KAAIVCYDLT DSSSFERAKF WVKELRSLEE GCQIYLCGTK SDLLEEDRRR RRVDFHDVQD YA Antigen sequence identity: Mouse 100%; Rat 100%.
RAB24 may be involved in autophagy-related processes.
Specifications
| RAB24 | |
| Polyclonal | |
| Unconjugated | |
| RAB24 | |
| 6530406O07Rik; rab-16; RAB24; RAB24, member RAS oncogene family; ras-related protein Rab-24 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 53917 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q969Q5 | |
| RAB24 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 82-143. Recombinant protein control fragment (Product #RP-109794). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction