missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB24 Monoclonal Antibody (OTI3H1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526188
This item is not returnable.
View return policy
Description
RAB24 Monoclonal Antibody for Western Blot, ICC/IF
RAB24 may be involved in autophagy-related processes.
Specifications
| RAB24 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P35290, Q969Q5 | |
| RAB24 | |
| Full length human recombinant protein of RAB24 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Canine | |
| Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| OTI3H1 | |
| Unconjugated | |
| RAB24 | |
| 6530406O07Rik; rab-16; RAB24; RAB24, member RAS oncogene family; ras-related protein Rab-24 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 19336, 479277, 53917 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction