missing translation for 'onlineSavingsMsg'
Learn More

RAB23 Antibody (OTI2F3) [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30687636 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30687636 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687636 Supplier Novus Biologicals Supplier No. NBP344767JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RAB23 Monoclonal antibody specifically detects RAB23 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB23
Applications Western Blot
Classification Monoclonal
Clone OTI2F3
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias DKFZp781H0695, HSPC137, MGC8900, RAB family small GTP binding protein RAB 23, RAB23, member RAS oncogene family, ras-related protein Rab-23
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-234 of human RAB23 (NP_899050) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51715
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.