Learn More
Invitrogen™ RAB22A Recombinant Rabbit Monoclonal Antibody (24GB1000)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557199
 View more versions of this product
Product Code. 30275731
  • £434.00 / 100µL
Description
RAB22A Recombinant Monoclonal Antibody for Western Blot, Flow
The protein encoded by this gene is a member of the RAB family of small GTPases. The GTP-bound form of the encoded protein has been shown to interact with early-endosomal antigen 1.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RAB22A
Recombinant Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P35285, Q9UL26
RAB22A
A synthesized peptide derived from human RAB22A (aa 5-54).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1000
Unconjugated
RAB22A
3732413A17Rik; AI662177; AW319644; AW550514; E130120E14Rik; GTP-binding protein RAB22A; im:7136651; MGC16770; rab-14; Rab22; Rab-22; RAB22, member RAS oncogene family; Rab22a; Rab22a protein; RAB22A, member RAS oncogene family; RAS-related protein RAB-22A; zgc:101631
Rabbit
Affinity chromatography
RUO
19334, 366265, 57403
-20°C