missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB22A Recombinant Rabbit Monoclonal Antibody (24GB1000)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557200
This item is not returnable.
View return policy
Description
RAB22A Recombinant Monoclonal Antibody for Western Blot, Flow
The protein encoded by this gene is a member of the RAB family of small GTPases. The GTP-bound form of the encoded protein has been shown to interact with early-endosomal antigen 1.
Specifications
| RAB22A | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P35285, Q9UL26 | |
| RAB22A | |
| A synthesized peptide derived from human RAB22A (aa 5-54). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1000 | |
| Unconjugated | |
| RAB22A | |
| 3732413A17Rik; AI662177; AW319644; AW550514; E130120E14Rik; GTP-binding protein RAB22A; im:7136651; MGC16770; rab-14; Rab22; Rab-22; RAB22, member RAS oncogene family; Rab22a; Rab22a protein; RAB22A, member RAS oncogene family; RAS-related protein RAB-22A; zgc:101631 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19334, 366265, 57403 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction