missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101822
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RAB20.
Plays a role in apical endocytosis/recycling. Plays a role in the maturation and acidification of phagosomes that engulf pathogens, such as S.aureus and M.tuberculosis. Plays a role in the fusion of phagosomes with lysosomes. [UniProt].
Specifications
| RAB20 | |
| Polyclonal | |
| Unconjugated | |
| Rab20 | |
| AA536966; D8Ertd350e; RAB20; RAB20, member RAS oncogene family; ras-related protein Rab-20 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55647, 689377 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NX57 | |
| Rab20 | |
| A synthesized peptide derived from human RAB20(Accession Q9NX57), corresponding to amino acid residues K114-Y164. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction